Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other COMT SIRNA Assays ›
          Gene Symbol:
          COMT
          Gene Name:
          catechol-O-methyltransferase
          Gene Aliases:
          HEL-S-98n
          Chromosome Location:
          Chr.22: 19941740 - 19969975 on Build GRCh38
          UniGene ID:
          Hs.370408
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s41
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: COMT
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000754.3 NP_000745.1 6 1234
          NM_001135161.1 NP_001128633.1 6 1192
          NM_001135162.1 NP_001128634.1 6 1209
          NM_007310.2 NP_009294.1 4 965
          XM_011529886.1 XP_011528188.1 6 1139
          XM_017028594.1 XP_016884083.1 6 1458
          XM_017028595.1 XP_016884084.1 6 1402
          GenBank mRNA AK130031.1 - 7 1015
          AK290440.1 - 6 1173
          BC000419.2 - 6 1163 2964400
          BC005867.2 - 6 1163 2964400
          BC011935.2 - 5 1078 3503220
          BC100018.1 - 6 1224 6163659
          FJ224345.1 - 6 1163
          M58525.1 - 6 1188
          M65212.1 - 5 1063
          M65213.1 - 1 235

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00241349_m1

          Panther Classification:

          Molecular Function -

          methyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          female pregnancy
          learning
          short-term memory
          estrogen metabolic process
          response to organic cyclic compound
          cellular response to phosphate starvation
          methylation
          response to lipopolysaccharide
          developmental process
          negative regulation of renal sodium excretion
          neurotransmitter catabolic process
          dopamine catabolic process
          response to drug
          negative regulation of dopamine metabolic process
          response to pain
          multicellular organismal reproductive process
          negative regulation of smooth muscle cell proliferation
          positive regulation of homocysteine metabolic process
          regulation of sensory perception of pain
          magnesium ion binding
          protein binding
          methyltransferase activity
          O-methyltransferase activity
          catechol O-methyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-dzzjg:80/100.66.77.174:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0