Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DLG3 SIRNA Assays ›
          Gene Symbol:
          DLG3
          Gene Name:
          discs large MAGUK scaffold protein 3
          Gene Aliases:
          MRX, MRX90, NEDLG, PPP1R82, SAP102, XLMR
          Chromosome Location:
          Chr.X: 70444850 - 70505490 on Build GRCh38
          UniGene ID:
          Hs.721586
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s4125
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DLG3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001166278.1 NP_001159750.1 12 1237
          NM_020730.2 NP_065781.1 14 1780
          NM_021120.3 NP_066943.2 19 2725
          XM_005262248.3 XP_005262305.1 14 1377
          XM_006724625.2 XP_006724688.1 20 2826
          XM_006724626.2 XP_006724689.1 20 2772
          XM_011530883.1 XP_011529185.1 19 2784
          XM_017029322.1 XP_016884811.1 13 1727
          XM_017029323.1 XP_016884812.1 14 1715
          XM_017029324.1 XP_016884813.1 13 1693
          XM_017029325.1 XP_016884814.1 12 3087
          XM_017029326.1 XP_016884815.1 12 1166
          XM_017029327.1 XP_016884816.1 11 1136
          GenBank mRNA AB033058.1 - 14 1778
          AB209328.1 - 19 2725
          AK303377.1 - 12 1237
          AK304020.1 - 14 1493
          AK315041.1 - 19 2664
          AK316518.1 - 14 1479
          BC093864.1 - 19 2426 7939709
          BC093866.1 - 19 2426 7939711
          U49089.1 - 19 2611

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_031639, XM_008773268, XM_006257081, XM_006257083, XM_006257085, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001177778, NM_001177779, NM_001177780, NM_016747, NM_001290402, and others.

          Matching Taqman Gene Expression Assay:


          Hs00221664_m1

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          establishment of planar polarity
          chemical synaptic transmission
          nervous system development
          negative regulation of cell proliferation
          negative regulation of phosphatase activity
          ion transmembrane transport
          receptor clustering
          establishment or maintenance of epithelial cell apical/basal polarity
          GMP metabolic process
          GDP metabolic process
          receptor localization to synapse
          guanylate kinase activity
          protein binding
          protein C-terminus binding
          ligand-gated ion channel activity
          kinase binding
          phosphatase binding
          protein phosphatase binding
          PDZ domain binding
          ubiquitin protein ligase binding
          ionotropic glutamate receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-qwrcb:80/100.66.75.127:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0