Search
Search

EGLN3| Gene Symbol: EGLN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308103.1 | NP_001295032.1 | 3, 4 | 411 | ||
| NM_022073.3 | NP_071356.1 | 3, 4 | 945 | |||
| GenBank mRNA | AJ310545.1 | - | 3, 4 | 945 | ||
| AK025273.1 | - | 3, 4 | 945 | |||
| AK123350.1 | - | 4, 5 | 774 | |||
| AK225473.1 | - | 3, 4 | 945 | |||
| BC010992.2 | - | 3, 4 | 901 | 3948676 | ||
| BC064924.1 | - | 3, 4 | 905 | 6141425 | ||
| BC102030.1 | - | 3, 4 | 411 | 40032049 | ||
| BC105938.1 | - | 3, 4 | 411 | 40032045 | ||
| BC105939.1 | - | 3, 4 | 693 | 40032051 | ||
| BC111057.1 | - | 3, 4 | 928 | 5747720 | ||
| BX346941.2 | - | 2, 3 | 268 | |||
| DQ975379.1 | - | 3, 4 | 345 | |||