Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRDMT1 SIRNA Assays ›
Gene Symbol:
TRDMT1
Gene Name:
tRNA aspartic acid methyltransferase 1
Gene Aliases:
DMNT2, DNMT2, MHSAIIP, PUMET, RNMT1
Chromosome Location:
Chr.10: 17136785 - 17202071 on Build GRCh38
UniGene ID:
Hs.351665
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s4219
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRDMT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321006.1
NP_001307935.1
2
555
NM_001321007.1
NP_001307936.1
2
555
NM_004412.6
NP_004403.1
2
555
XM_005252372.4
XP_005252429.1
2
550
XM_005252374.4
XP_005252431.1
2
550
XM_005252375.4
XP_005252432.1
2
550
XM_005252378.3
XP_005252435.1
2
151
XM_006717388.3
XP_006717451.1
2
550
XM_011519343.2
XP_011517645.1
2
550
XM_011519344.2
XP_011517646.1
2
550
XM_011519345.2
XP_011517647.1
2
963
XM_017015800.1
XP_016871289.1
2
550
XM_017015801.1
XP_016871290.1
2
550
XM_017015802.1
XP_016871291.1
1
54
XM_017015803.1
XP_016871292.1
2
147
XM_017015804.1
XP_016871293.1
2
154
XM_017015806.1
XP_016871295.1
2
151
XM_017015807.1
XP_016871296.1
2
172
XM_017015808.1
XP_016871297.1
2
149
XM_017015809.1
XP_016871298.1
2
550
XM_017015810.1
XP_016871299.1
2
151
GenBank mRNA
AF012128.1
-
2
125
AF045888.1
-
2
124
AF329940.1
-
1
64
AF329941.1
-
1
64
AF329942.1
-
1
64
AF329943.1
-
1
64
AF329944.1
-
1
64
AJ223333.1
-
2
166
AK291417.1
-
2
124
AK299027.1
-
2
335
AK303428.1
-
2
141
BC047733.1
-
2
125
6473993
BX537961.1
-
2
145
CR450316.1
-
2
118
CV816165.1
-
2
538
DC411638.1
-
2
555
HQ637546.1
-
2
118
HQ637547.1
-
2
118
HQ637548.1
-
2
118
HQ637549.1
-
2
118
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
RNA processing factor
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
response to amphetamine
tRNA modification
tRNA methylation
C-5 methylation of cytosine
RNA binding
DNA (cytosine-5-)-methyltransferase activity
tRNA methyltransferase activity
tRNA (cytosine-5-)-methyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0