Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AGAP6 SIRNA Assays ›
          Gene Symbol:
          AGAP6 AGAP12P AGAP4 AGAP5 AGAP9 CCNYL2 LOC107984026
          Gene Name:
          ArfGAP with GTPase domain, ankyrin repeat and PH domain 6
          ArfGAP with GTPase domain, ankyrin repeat and PH domain 12, pseudogene
          ArfGAP with GTPase domain, ankyrin repeat and PH domain 4
          ArfGAP with GTPase domain, ankyrin repeat and PH domain 5
          ArfGAP with GTPase domain, ankyrin repeat and PH domain 9
          cyclin Y-like 2 (pseudogene)
          arf-GAP with GTPase, ANK repeat and PH domain-containing protein 4-like
          Gene Aliases:
          AGAP-6, CTGLF3, bA324H6.1
          CTGLF12P
          AGAP-4, AGAP-8, AGAP8, CTGLF1, CTGLF5, MRIP2
          CTGLF2
          AGAP-9, CTGLF6, bA301J7.2
          C10orf21, CCNYL2P, bA178A10.2
          -
          Chromosome Location:
          Chr.10: 49981196 - 50010507 on Build GRCh38
          Chromosome Location:
          Chr.10: 48010132 - 48031640 on Build GRCh38
          Chromosome Location:
          Chr.10: 45825592 - 45858529 on Build GRCh38
          Chromosome Location:
          Chr.10: 73674275 - 73697796 on Build GRCh38
          Chromosome Location:
          Chr.10: 47501854 - 47523638 on Build GRCh38
          Chromosome Location:
          Chr.10: 42408174 - 42472240 on Build GRCh38
          Chromosome Location:
          Chr.10: 46335552 - 46838268 on Build GRCh38
          UniGene ID:
          Hs.536158
          Hs.645213
          Hs.314437
          Hs.651147
          Hs.645346
          Hs.568048
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s42269

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: AGAP6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001077665.2 NP_001071133.2 5 843
          XM_011539815.2 XP_011538117.1 2 68
          XM_017016268.1 XP_016871757.1 5 564
          XM_017016269.1 XP_016871758.1 5 564
          XM_017016270.1 XP_016871759.1 4 798
          XM_017016271.1 XP_016871760.1 7 566
          XM_017016272.1 XP_016871761.1 6 496
          XM_017016273.1 XP_016871762.1 5 855
          Gene Symbol: AGAP12P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_029396.2 5 487
          Gene Symbol: AGAP4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001276343.2 NP_001263272.2 5 571
          NM_001291379.1 NP_001278308.1 7 763
          NM_133446.3 NP_597703.2 4 502
          XM_017015664.1 XP_016871153.1 7 4433
          XM_017015665.1 XP_016871154.1 8 4301
          XM_017015666.1 XP_016871155.1 6 1336
          XM_017015667.1 XP_016871156.1 7 606
          XM_017015668.1 XP_016871157.1 4 222
          XM_017015669.1 XP_016871158.1 4 175
          GenBank mRNA AF411132.1 - 4 502
          BC171820.1 - 5 556
          Gene Symbol: AGAP5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001144000.1 NP_001137472.1 5 486
          Gene Symbol: AGAP9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001190810.1 NP_001177739.1 5 557
          GenBank mRNA AK160375.1 - 6 3533
          BC171845.1 - 5 557
          Gene Symbol: CCNYL2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_103829.1 4 747
          Gene Symbol: LOC107984026
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_017017029.1 XP_016872518.1 7 988
          XM_017017030.1 XP_016872519.1 6 919
          XM_017017031.1 XP_016872520.1 6 919
          XM_017017032.1 XP_016872521.1 8 1019
          XM_017017033.1 XP_016872522.1 9 782
          XM_017017034.1 XP_016872523.1 8 687
          XM_017017035.1 XP_016872524.1 4 200
          XM_017017036.1 XP_016872525.1 2 361
          XM_017017039.1 XP_016872528.1 4 219
          XM_017017040.1 XP_016872529.1 4 183

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of GTPase activity
          regulation of cyclin-dependent protein serine/threonine kinase activity
          GTPase activator activity
          metal ion binding
          protein kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-g6tbm:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0