Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RAVER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133452.2 | NP_597709.2 | 9 | 1659 | ||
XM_011527671.1 | XP_011525973.1 | 9 | 1653 | |||
GenBank mRNA | AB075858.1 | - | 6 | 2762 | ||
BC037565.1 | - | 7 | 996 |