Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: OR1M1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004456.1 | NP_001004456.1 | 1 | 825 | ||
GenBank mRNA | BC137551.1 | - | 1 | 891 | 9021551 | |
BC137555.1 | - | 1 | 891 | 9021555 |