Search
Search

FCRLB| Gene Symbol: FCRLB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002901.3 | NP_001002901.1 | 5 | 457 | ||
| NM_001288829.1 | NP_001275758.1 | 3 | 243 | |||
| NM_001288830.1 | NP_001275759.1 | 2 | 222 | |||
| NM_001288831.1 | NP_001275760.1 | 3 | 243 | |||
| NM_001288832.1 | NP_001275761.1 | 2 | 222 | |||
| NM_001320241.1 | NP_001307170.1 | 4 | 271 | |||
| GenBank mRNA | AK290733.1 | - | 4 | 271 | ||
| AY670683.1 | - | 3 | 238 | |||
| AY670684.1 | - | 3 | 238 | |||
| AY670685.1 | - | 2 | 217 | |||
| AY670686.1 | - | 3 | 238 | |||
| AY670687.1 | - | 2 | 217 | |||
| AY786314.1 | - | 3 | 223 | |||
| BX364259.2 | - | 4 | 417 | |||
| DQ021957.1 | - | 5 | 457 | |||