Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DUT SIRNA Assays ›
          Gene Symbol:
          DUT
          Gene Name:
          deoxyuridine triphosphatase
          Gene Aliases:
          dUTPase
          Chromosome Location:
          Chr.15: 48331018 - 48343373 on Build GRCh38
          UniGene ID:
          Hs.527980
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s4390
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DUT
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001025248.1 NP_001020419.1 6 757
          NM_001025249.1 NP_001020420.1 6 441
          NM_001948.3 NP_001939.1 5 485
          XM_011521315.2 XP_011519617.1 6 477
          GenBank mRNA AB049113.2 - 5 420
          AK000629.1 - 6 437
          AK291515.1 - 5 474
          AK298464.1 - 6 733
          AK312122.1 - 5 453
          BC033645.1 - 5 513 5578877
          BC070339.1 - 6 741 3998021
          BC110377.1 - 6 736 5728970
          BG505144.1 - 6 457
          BG532018.1 - 488
          CR541720.1 - 5 401
          CR541781.1 - 5 401
          L11877.1 - 5 421
          M89913.1 - 5 419
          U31930.1 - 5 430
          U62891.1 - 5 432
          U90223.1 - 6 727

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00798995_s1

          Panther Classification:

          Molecular Function -

          hydrolase phosphatase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleobase-containing compound metabolic process
          dUMP biosynthetic process
          DNA replication
          dUTP catabolic process
          pyrimidine nucleoside biosynthetic process
          magnesium ion binding
          dUTP diphosphatase activity
          protein binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0