Search
Search

DYRK1A| Gene Symbol: DYRK1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001396.3 | NP_001387.2 | 9 | 1422 | ||
| NM_101395.2 | NP_567824.1 | 11 | 1596 | |||
| NM_130436.2 | NP_569120.1 | 9 | 2790 | |||
| NM_130438.2 | NP_569122.1 | 9 | 1423 | |||
| XM_005260933.4 | XP_005260990.1 | 5 | 749 | |||
| XM_006723976.3 | XP_006724039.1 | 10 | 1494 | |||
| XM_006723977.3 | XP_006724040.1 | 10 | 1819 | |||
| XM_006723978.3 | XP_006724041.1 | 10 | 2548 | |||
| XM_006723979.3 | XP_006724042.1 | 10 | 1654 | |||
| XM_011529482.2 | XP_011527784.1 | 9 | 1516 | |||
| XM_011529483.2 | XP_011527785.1 | 10 | 4362 | |||
| XM_011529484.2 | XP_011527786.1 | 9 | 1489 | |||
| XM_011529485.2 | XP_011527787.1 | 9 | 2005 | |||
| XM_017028284.1 | XP_016883773.1 | 10 | 1453 | |||
| XM_017028285.1 | XP_016883774.1 | 10 | 1699 | |||
| XM_017028286.1 | XP_016883775.1 | 12 | 1696 | |||
| GenBank mRNA | AF108830.1 | - | 11 | 1596 | ||
| AK301752.1 | - | 5 | 762 | |||
| D85759.1 | - | 9 | 1388 | |||
| D86550.1 | - | 9 | 2793 | |||
| U52373.1 | - | 9 | 1422 | |||
| U58496.1 | - | 9 | 1382 | |||