Search
Search

SUN5| Gene Symbol: SUN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080675.3 | NP_542406.2 | 7 | 499 | ||
| XM_011528573.1 | XP_011526875.1 | 8 | 583 | |||
| XM_011528574.1 | XP_011526876.1 | 6 | 439 | |||
| XM_011528575.1 | XP_011526877.1 | 4 | 218 | |||
| XM_011528576.1 | XP_011526878.1 | 8 | 584 | |||
| GenBank mRNA | AF401350.1 | - | 7 | 499 | ||
| BC026118.2 | - | 7 | 499 | 4824412 | ||
| BC029528.1 | - | 7 | 492 | 5271065 | ||