Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080826.1 | NP_543016.1 | 3 | 623 | ||
XM_017027680.1 | XP_016883169.1 | 3 | 623 | |||
GenBank mRNA | AK307409.1 | - | 1 | 62 | ||
CR982468.1 | - | 2 | 404 | |||
DB202450.1 | - | 1 | 62 |