Search
Search

EFCAB3| Gene Symbol: EFCAB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144933.1 | NP_001138405.1 | 11 | 1201 | ||
| NM_173503.3 | NP_775774.1 | 9 | 1052 | |||
| XM_011524380.1 | XP_011522682.1 | 9 | 1051 | |||
| XM_011524381.1 | XP_011522683.1 | 9 | 1019 | |||
| GenBank mRNA | AK098684.1 | - | 9 | 1053 | ||
| BC101752.1 | - | 9 | 1025 | 8069258 | ||
| BC101778.1 | - | 9 | 1025 | 8069284 | ||