Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TMEM199 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152464.2 | NP_689677.1 | 3, 4 | 464 | ||
GenBank mRNA | AK291475.1 | - | 3, 4 | 387 | ||
AK311363.1 | - | 346 | ||||
AY074907.1 | - | 3, 4 | 374 | |||
BC033113.1 | - | 3, 4 | 381 | 5429060 | ||
DC327155.1 | - | 3, 4 | 464 |