Search
Search

CAPN12| Gene Symbol: CAPN12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144691.4 | NP_653292.2 | 5 | 917 | ||
| XM_017026355.1 | XP_016881844.1 | 5 | 917 | |||
| XM_017026356.1 | XP_016881845.1 | 5 | 917 | |||
| XM_017026357.1 | XP_016881846.1 | 5 | 917 | |||
| XM_017026358.1 | XP_016881847.1 | 5 | 917 | |||
| XM_017026359.1 | XP_016881848.1 | 5 | 917 | |||
| XM_017026360.1 | XP_016881849.1 | 5 | 917 | |||
| XM_017026361.1 | XP_016881850.1 | 5 | 917 | |||
| XM_017026362.1 | XP_016881851.1 | 5 | 917 | |||
| XM_017026363.1 | XP_016881852.1 | 5 | 917 | |||
| XM_017026365.1 | XP_016881854.1 | 5 | 917 | |||
| GenBank mRNA | AK127398.1 | - | 5 | 917 | ||