Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCA5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152505.3 | NP_689718.1 | 4 | 425 | ||
XM_005260926.1 | XP_005260983.1 | 4 | 383 | |||
XM_006723967.1 | XP_006724030.1 | 3 | 428 | |||
XM_011529458.1 | XP_011527760.1 | 5 | 834 | |||
XM_011529459.1 | XP_011527761.1 | 5 | 493 | |||
XM_011529460.1 | XP_011527762.1 | 6 | 633 | |||
XM_011529461.1 | XP_011527763.1 | 6 | 740 | |||
XM_011529462.1 | XP_011527764.1 | 4 | 383 | |||
XM_011529463.2 | XP_011527765.1 | 4 | 436 | |||
XM_011529464.1 | XP_011527766.1 | 5 | 462 | |||
XM_011529465.1 | XP_011527767.1 | 4 | 381 | |||
XM_011529466.1 | XP_011527768.1 | 4 | 619 | |||
XM_011529467.1 | XP_011527769.1 | 5 | 697 | |||
XM_011529468.1 | XP_011527770.1 | 4 | 339 | |||
XM_011529469.2 | XP_011527771.1 | 4 | 726 | |||
XM_011529470.1 | XP_011527772.1 | 3 | 222 | |||
XM_011529471.1 | XP_011527773.1 | 3 | 428 | |||
GenBank mRNA | BC031059.2 | - | 4 | 426 | 5271223 | |
BC043006.1 | - | 4 | 642 | 5297703 | ||
BX647793.1 | - | 3 | 460 | |||
BX648744.1 | - | 1 | 53 |