Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
CDMO and Clinical Trial Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKAR SIRNA Assays ›
Gene Symbol:
ANKAR
Gene Name:
ankyrin and armadillo repeat containing
Gene Aliases:
-
Chromosome Location:
Chr.2: 189674292 - 189763059 on Build GRCh38
UniGene ID:
Hs.60772
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s45477
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKAR
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_144708.3
NP_653309.3
20
3868
XM_011510673.2
XP_011508975.1
20
4387
XM_011510674.1
XP_011508976.1
20
4354
XM_011510675.2
XP_011508977.1
19
4282
XM_011510676.2
XP_011508978.1
20
4387
XM_011510677.2
XP_011508979.1
20
4387
XM_011510679.2
XP_011508981.1
18
3958
XM_011510680.1
XP_011508982.1
20
4387
XM_011510681.1
XP_011508983.1
20
4387
XM_011510682.2
XP_011508984.1
20
4387
XM_011510685.2
XP_011508987.1
20
3943
XM_011510686.2
XP_011508988.1
16
3880
XM_011510687.2
XP_011508989.1
19
3751
XM_011510688.1
XP_011508990.1
18
4110
XM_011510691.2
XP_011508993.1
16
2573
XM_011510692.2
XP_011508994.1
18
3587
XM_011510693.2
XP_011508995.1
17
3483
XM_011510694.2
XP_011508996.1
16
3302
XM_017003413.1
XP_016858902.1
20
4387
XM_017003415.1
XP_016858904.1
15
3082
XM_017003416.1
XP_016858905.1
17
3494
XM_017003418.1
XP_016858907.1
15
3197
XM_017003419.1
XP_016858908.1
15
3164
XM_017003420.1
XP_016858909.1
15
3100
GenBank mRNA
AJ549812.1
-
19
3567
AJ605761.1
-
8
1036
AK058144.1
-
8
1220
AK126073.1
-
18
3638
BC037215.1
-
14
2857
BC044907.1
-
16
3361
4836761
BC047413.1
-
23
3432
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-55874ff697-bg9ld:80/10.239.161.145:80.
git-commit: a2f5e26549390d543f5d2d1fbad8092df65fbda4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.10.0-2024.06.23-1.0