Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GPR155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033045.3 | NP_001028217.1 | 6 | 1467 | ||
NM_001267050.1 | NP_001253979.1 | 6 | 1364 | |||
NM_001267051.1 | NP_001253980.1 | 5 | 1318 | |||
NM_152529.6 | NP_689742.4 | 5 | 1318 | |||
XM_017003485.1 | XP_016858974.1 | 7 | 1509 | |||
XM_017003486.1 | XP_016858975.1 | 5 | 1370 | |||
XM_017003487.1 | XP_016858976.1 | 6 | 1371 | |||
GenBank mRNA | AK056381.1 | - | 3 | 566 | ||
AK091200.1 | - | 5 | 1259 | |||
AK307896.1 | - | 5 | 1310 | |||
AK315125.1 | - | 5 | 1259 | |||
AL834154.1 | - | 6 | 1416 | |||
BC028730.1 | - | 6 | 1413 | 4813561 | ||
BC035037.1 | - | 6 | 1375 | 5260361 | ||
BC036681.1 | - | 5 | 1264 | |||
BX537947.1 | - | 5 | 1311 |