Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCDC138 SIRNA Assays ›
Gene Symbol:
CCDC138
Gene Name:
coiled-coil domain containing 138
Gene Aliases:
-
Chromosome Location:
Chr.2: 108786730 - 108885485 on Build GRCh38
UniGene ID:
Hs.362702
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s46551
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCDC138
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001303105.1
NP_001290034.1
9
1020
NM_001303106.1
NP_001290035.1
9
1015
NM_144978.2
NP_659415.1
9
1020
NR_130121.1
9
1140
XM_006712344.2
XP_006712407.1
9
1005
XM_006712345.3
XP_006712408.1
9
1005
XM_011510757.2
XP_011509059.1
9
1002
XM_011510758.2
XP_011509060.1
8
982
XM_011510760.2
XP_011509062.1
9
1005
XM_011510761.2
XP_011509063.1
9
1005
XM_011510762.2
XP_011509064.1
9
1005
XM_017003492.1
XP_016858981.1
9
1015
XM_017003493.1
XP_016858982.1
8
978
XM_017003494.1
XP_016858983.1
9
1018
XM_017003495.1
XP_016858984.1
9
1015
XM_017003496.1
XP_016858985.1
9
1002
XM_017003497.1
XP_016858986.1
9
1015
XM_017003498.1
XP_016858987.1
9
1005
XM_017003499.1
XP_016858988.1
9
1018
XM_017003500.1
XP_016858989.1
8
977
XM_017003501.1
XP_016858990.1
9
914
XM_017003502.1
XP_016858991.1
9
965
XM_017003503.1
XP_016858992.1
9
904
XM_017003504.1
XP_016858993.1
8
919
GenBank mRNA
AK057307.1
-
9
1007
AK125199.1
-
9
1129
BC015847.1
-
9
1001
BC038421.1
-
9
976
BC050579.1
-
9
1013
5764613
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-sjvkz:80/100.66.74.89:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0