Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: DCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242377.1 | NP_001229306.1 | 5 | 743 | ||
NM_152624.5 | NP_689837.2 | 5 | 743 | |||
NR_038352.1 | 3 | 463 | ||||
XM_017009163.1 | XP_016864652.1 | 6 | 835 | |||
GenBank mRNA | AK090564.1 | - | 5 | 658 | ||
AK296361.1 | - | 3 | 393 | |||
AY135173.1 | - | 5 | 717 | |||
AY146650.1 | - | 5 | 717 | |||
BC045596.1 | - | 1 | 40 | |||
BC064593.1 | - | 5 | 649 | 5757231 | ||
BG720562.1 | - | 3 | 469 | |||
BX648838.1 | - | 2 | 155 |