Search
Search

ADAMTS17| Gene Symbol: ADAMTS17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139057.3 | NP_620688.2 | 22 | 3290 | ||
| XM_005254872.3 | XP_005254929.1 | 23 | 4287 | |||
| XM_011521312.2 | XP_011519614.1 | 24 | 4356 | |||
| XM_017021973.1 | XP_016877462.1 | 25 | 4489 | |||
| XM_017021975.1 | XP_016877464.1 | 24 | 4420 | |||
| XM_017021976.1 | XP_016877465.1 | 24 | 3201 | |||
| XM_017021978.1 | XP_016877467.1 | 18 | 5747 | |||
| XM_017021979.1 | XP_016877468.1 | 17 | 2197 | |||
| XM_017021980.1 | XP_016877469.1 | 17 | 2407 | |||
| XM_017021982.1 | XP_016877471.1 | 15 | 2017 | |||
| XM_017021983.1 | XP_016877472.1 | 13 | 1746 | |||
| GenBank mRNA | AJ315735.1 | - | 22 | 3207 | ||
| AK057331.1 | - | 2 | 284 | |||