Search
Search

ADAMTS18| Gene Symbol: ADAMTS18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001326358.1 | NP_001313287.1 | 3 | 645 | ||
| NM_199355.3 | NP_955387.1 | 3 | 645 | |||
| XM_011522923.1 | XP_011521225.1 | 1 | 52 | |||
| XM_011522924.1 | XP_011521226.1 | 1 | 48 | |||
| XM_017022988.1 | XP_016878477.1 | 1 | 48 | |||
| GenBank mRNA | AJ311903.1 | - | 3 | 308 | ||
| AK131403.1 | - | 2 | 152 | |||
| AK293831.1 | - | 3 | 565 | |||
| BC063283.1 | - | 3 | 645 | 30343625 | ||