Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DMTN SIRNA Assays ›
          Gene Symbol:
          DMTN
          Gene Name:
          dematin actin binding protein
          Gene Aliases:
          DMT, EPB49
          Chromosome Location:
          Chr.8: 22048955 - 22082527 on Build GRCh38
          UniGene ID:
          Hs.106124
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s4715

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DMTN
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001114135.4 NP_001107607.1 9, 10 1003
          NM_001114136.2 NP_001107608.1 9, 10 959
          NM_001114137.3 NP_001107609.1 9, 10 1133
          NM_001114138.2 NP_001107610.1 9, 10 1120
          NM_001114139.3 NP_001107611.1 8, 9 1017
          NM_001302816.2 NP_001289745.1 9, 10 1253
          NM_001302817.2 NP_001289746.1 7, 8 972
          NM_001323378.1 NP_001310307.1 9, 10 1092
          NM_001323380.1 NP_001310309.1 9, 10 999
          NM_001323381.1 NP_001310310.1 8, 9 1108
          NM_001323382.1 NP_001310311.1 9, 10 1038
          NM_001323383.1 NP_001310312.1 9, 10 959
          NM_001323384.1 NP_001310313.1 9, 10 1092
          NM_001323385.1 NP_001310314.1 8, 9 1108
          NM_001323387.1 NP_001310316.1 8, 9 1057
          NM_001323388.1 NP_001310317.1 7, 8 839
          NM_001323389.1 NP_001310318.1 8, 9 1047
          NM_001323390.1 NP_001310319.1 8, 9 914
          NM_001323391.1 NP_001310320.1 8, 9 1017
          NM_001323392.1 NP_001310321.1 8, 9 1178
          NM_001323393.1 NP_001310322.1 8, 9 884
          NM_001323394.1 NP_001310323.1 8, 9 1045
          NM_001323395.1 NP_001310324.1 8, 9 924
          NM_001323396.1 NP_001310325.1 8, 9 928
          NM_001323397.1 NP_001310326.1 8, 9 1096
          NM_001323398.1 NP_001310327.1 7, 8 972
          NM_001323399.1 NP_001310328.1 7, 8 839
          NM_001323400.1 NP_001310329.1 6, 7 988
          NM_001323401.1 NP_001310330.1 7, 8 1012
          NM_001978.4 NP_001969.2 9, 10 1253
          XM_005273432.1 XP_005273489.1 9, 10 985
          XM_005273433.1 XP_005273490.1 9, 10 917
          XM_005273442.1 XP_005273499.1 8, 9 769
          XM_011544435.1 XP_011542737.1 8, 9 1178
          XM_017013189.1 XP_016868678.1 9, 10 926
          XM_017013190.1 XP_016868679.1 8, 9 1178
          XM_017013191.1 XP_016868680.1 8, 9 932
          XM_017013192.1 XP_016868681.1 8, 9 813
          XM_017013193.1 XP_016868682.1 7, 8 945
          XM_017013194.1 XP_016868683.1 7, 8 812
          XM_017013195.1 XP_016868684.1 8, 9 1178
          XM_017013196.1 XP_016868685.1 8, 9 781
          XM_017013197.1 XP_016868686.1 8, 9 1012
          XM_017013198.1 XP_016868687.1 8, 9 854
          GenBank mRNA AB209718.1 - 5, 6 479
          AK055842.1 - 9, 10 917
          AK091581.1 - 8, 9 932
          AK289650.1 - 9, 10 885
          AK295452.1 - 7, 8 858
          BC006318.2 - 9, 10 1002 4125804
          BC017445.1 - 9, 10 1002 3162732
          BC052805.1 - 9, 10 1043 6269658
          BT007396.1 - 8, 9 730
          L19713.1 - 9, 10 1180
          U28389.1 - 9, 10 1180

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001108385, XM_008770831, XM_006252279, XM_006252281, XM_008770830, and others.

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein complex assembly
          cytoskeleton organization
          regulation of cell shape
          regulation of lamellipodium assembly
          positive regulation of fibroblast migration
          negative regulation of peptidyl-threonine phosphorylation
          negative regulation of cell-substrate adhesion
          actin cytoskeleton organization
          positive regulation of blood coagulation
          regulation of actin cytoskeleton organization
          negative regulation of peptidyl-serine phosphorylation
          calcium-mediated signaling using intracellular calcium source
          calcium-mediated signaling using extracellular calcium source
          erythrocyte development
          negative regulation of peptidyl-tyrosine phosphorylation
          actin filament bundle assembly
          regulation of filopodium assembly
          actin filament capping
          negative regulation of focal adhesion assembly
          transmembrane transport
          protein secretion by platelet
          cellular response to calcium ion
          cellular response to cAMP
          positive regulation of wound healing
          negative regulation of protein targeting to membrane
          actin filament reorganization
          negative regulation of substrate adhesion-dependent cell spreading
          positive regulation of substrate adhesion-dependent cell spreading
          positive regulation of platelet aggregation
          positive regulation of integrin-mediated signaling pathway
          actin binding
          receptor binding
          protein binding
          spectrin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0