Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012758.2 | NP_001012776.1 | 4 | 428 | ||
XM_011509260.2 | XP_011507562.1 | 2 | 866 | |||
XM_011509262.1 | XP_011507564.1 | 4 | 1076 | |||
XM_017000560.1 | XP_016856049.1 | 3 | 1050 | |||
GenBank mRNA | AK298083.1 | - | 3 | 568 | ||
BG820863.1 | - | 4 | 428 |