Search
Search

TMEM154| Gene Symbol: TMEM154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152680.2 | NP_689893.1 | 5 | 656 | ||
| GenBank mRNA | AK056590.1 | - | 5 | 530 | ||
| AK296694.1 | - | 5 | 481 | |||
| BC019600.1 | - | 4 | 259 | |||
| BC060775.1 | - | 5 | 428 | 30336271 | ||