Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLC36A1 SIRNA Assays ›
          Gene Symbol:
          SLC36A1
          Gene Name:
          solute carrier family 36 member 1
          Gene Aliases:
          Dct1, LYAAT1, PAT1, TRAMD3
          Chromosome Location:
          Chr.5: 151411344 - 151556318 on Build GRCh38
          UniGene ID:
          Hs.269004
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s47578

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SLC36A1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001308150.1 NP_001295079.1 2, 3 367
          NM_001308151.1 NP_001295080.1 2, 3 337
          NM_078483.3 NP_510968.2 2, 3 367
          XM_005268386.1 XP_005268443.1 2, 3 337
          XM_006714759.3 XP_006714822.1 2, 3 370
          XM_011537580.2 XP_011535882.1 2, 3 1057
          XM_011537581.1 XP_011535883.1 3, 4 485
          XM_011537583.2 XP_011535885.1 3, 4 4922
          XM_011537584.2 XP_011535886.1 2, 3 576
          XM_011537585.1 XP_011535887.1 3, 4 432
          XM_011537586.2 XP_011535888.1 2, 3 1180
          XM_011537587.2 XP_011535889.1 2, 3 436
          XM_011537588.2 XP_011535890.1 2, 3 280
          XM_011537589.2 XP_011535891.1 2, 3 1188
          XM_011537590.1 XP_011535892.1 2, 3 195
          XM_011537591.1 XP_011535893.1 2, 3 170
          XM_011537592.2 XP_011535894.1 2, 3 174
          XM_011537593.2 XP_011535895.1 2, 3 371
          XM_011537594.1 XP_011535896.1 3, 4 485
          XM_011537595.2 XP_011535897.2 2, 3 1063
          XM_011537596.2 XP_011535898.2 1, 2 149
          XM_017009216.1 XP_016864705.1 2, 3 263
          XM_017009217.1 XP_016864706.1 2, 3 367
          XM_017009218.1 XP_016864707.1 1, 2 149
          XM_017009219.1 XP_016864708.1 2, 3 371
          XM_017009220.1 XP_016864709.1 2, 3 337
          GenBank mRNA AF516142.1 - 1, 2 159
          AK057340.1 - 2, 3 337
          AK309323.1 - 2, 3 339
          AY162213.1 - 2, 3 272
          AY227111.1 - 3, 4 287
          AY227113.1 - 2, 3 243
          BC136437.1 - 2, 3 238 9020426
          BC136438.1 - 2, 3 238 9020427
          BM787446.1 - 2, 3 370
          BX537963.1 - 2, 3 364
          GU296670.1 - 1, 2 144

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          amino acid transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          ion transport
          amino acid transport
          L-alanine transport
          glycine transport
          proline transport
          proline transmembrane transport
          hydrogen ion transmembrane transport
          hydrogen:amino acid symporter activity
          hydrogen ion transmembrane transporter activity
          amino acid transmembrane transporter activity
          L-alanine transmembrane transporter activity
          glycine transmembrane transporter activity
          L-proline transmembrane transporter activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-nswnx:80/100.66.78.82:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0