Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: OR5M3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004742.2 | NP_001004742.2 | 1 | 665 | ||
GenBank mRNA | BC136982.1 | - | 1 | 665 | 9020974 | |
BC136983.1 | - | 1 | 665 | 9020975 |