Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EREG SIRNA Assays ›
          Gene Symbol:
          EREG
          Gene Name:
          epiregulin
          Gene Aliases:
          EPR, ER, Ep
          Chromosome Location:
          Chr.4: 74365143 - 74388760 on Build GRCh38
          UniGene ID:
          Hs.115263
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s4791

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: EREG
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001432.2 NP_001423.1 4 569
          GenBank mRNA AK314959.1 - 4 569
          BC136404.1 - 4 526 9020391
          BC136405.1 - 4 526 9020392
          CR541887.1 - 4 403
          D30783.1 - 4 569

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          angiogenesis
          ovarian cumulus expansion
          oocyte maturation
          positive regulation of cytokine production
          female meiotic division
          epidermal growth factor receptor signaling pathway
          cell-cell signaling
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          mRNA transcription
          anatomical structure morphogenesis
          organ morphogenesis
          regulation of phosphatidylinositol 3-kinase signaling
          peptidyl-tyrosine phosphorylation
          cytokine-mediated signaling pathway
          keratinocyte differentiation
          ovulation
          ERBB2 signaling pathway
          wound healing
          positive regulation of cytokine biosynthetic process
          positive regulation of phosphorylation
          luteinizing hormone signaling pathway
          response to peptide hormone
          positive regulation of GTPase activity
          keratinocyte proliferation
          positive regulation of innate immune response
          positive regulation of interleukin-6 biosynthetic process
          positive regulation of DNA replication
          positive regulation of epidermal growth factor-activated receptor activity
          positive regulation of mitotic nuclear division
          positive regulation of protein kinase activity
          negative regulation of transcription, DNA-templated
          phosphatidylinositol phosphorylation
          phosphatidylinositol-mediated signaling
          positive regulation of fibroblast proliferation
          primary follicle stage
          positive regulation of smooth muscle cell proliferation
          negative regulation of epithelial cell proliferation
          negative regulation of smooth muscle cell differentiation
          positive regulation of cell division
          regulation of cell motility
          protein tyrosine kinase activity
          Ras guanyl-nucleotide exchange factor activity
          epidermal growth factor receptor binding
          protein binding
          growth factor activity
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0