Search Thermo Fisher Scientific
Gene Symbol: NT5DC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152729.2 | NP_689942.2 | 2, 3 | 280 | ||
XM_006715378.3 | XP_006715441.1 | 2, 3 | 283 | |||
GenBank mRNA | AK308337.1 | - | 2, 3 | 263 | ||
BC015138.1 | - | 1, 2 | 61 | |||
BC108699.1 | - | 2, 3 | 237 | 6061738 | ||
BC136832.1 | - | 2, 3 | 239 | 9020823 | ||
BC136841.1 | - | 2, 3 | 239 | 9020832 | ||
BM153683.1 | - | 2, 3 | 286 | |||
BM808222.1 | - | 269 |