Search
Search

ATP6V0D2| Gene Symbol: ATP6V0D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152565.1 | NP_689778.1 | 6, 7 | 890 | ||
| GenBank mRNA | AK096027.1 | - | 6, 7 | 890 | ||
| AY079172.1 | - | 6, 7 | 871 | |||
| BC065207.1 | - | 6, 7 | 896 | 5190344 | ||