Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STXBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178509.5 | NP_848604.3 | 5 | 420 | ||
XM_005257187.4 | XP_005257244.1 | 5 | 456 | |||
XM_006721797.3 | XP_006721860.1 | 5 | 454 | |||
XM_006721798.3 | XP_006721861.1 | 5 | 451 | |||
XM_017024410.1 | XP_016879899.1 | 5 | 457 | |||
XM_017024411.1 | XP_016879900.1 | 5 | 455 | |||
XM_017024412.1 | XP_016879901.1 | 5 | 454 | |||
XM_017024413.1 | XP_016879902.1 | 2 | 121 | |||
XM_017024414.1 | XP_016879903.1 | 5 | 456 | |||
XM_017024415.1 | XP_016879904.1 | 5 | 456 | |||
GenBank mRNA | AK122865.1 | - | 5 | 420 | ||
BC041485.2 | - | 4 | 257 | |||
BC125116.1 | - | 5 | 409 | 40118892 |