Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RALGAPA1 SIRNA Assays ›
Gene Symbol:
RALGAPA1
RALGAPA1P1
Gene Name:
Ral GTPase activating protein catalytic alpha subunit 1
Ral GTPase activating protein catalytic alpha subunit 1 pseudogene 1
Gene Aliases:
GARNL1, GRIPE, RalGAPalpha1, TULIP1, p240
GARNL2, GARNL2P, RALGAPA1P, bA235C23.1
Chromosome Location:
Chr.14: 35538352 - 35809304 on Build GRCh38
Chromosome Location:
Chr.9: 105519824 - 105527319 on Build GRCh38
UniGene ID:
Hs.113150
Hs.113150
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
s48467
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RALGAPA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001283043.1
NP_001269972.1
12
1865
NM_001283044.1
NP_001269973.1
12
1865
NM_014990.1
NP_055805.1
12
1865
NM_194301.2
NP_919277.2
12
1865
XM_005267491.3
XP_005267548.1
12
1943
XM_005267492.3
XP_005267549.1
12
1943
XM_005267493.3
XP_005267550.1
12
1943
XM_006720098.2
XP_006720161.1
12
1943
XM_006720099.2
XP_006720162.1
12
1943
XM_006720100.3
XP_006720163.1
12
1943
XM_006720101.2
XP_006720164.1
12
1943
XM_006720102.3
XP_006720165.1
12
1649
XM_006720103.2
XP_006720166.1
12
1943
XM_006720104.3
XP_006720167.1
12
1943
XM_011536615.2
XP_011534917.1
12
1943
XM_011536616.2
XP_011534918.1
12
1943
XM_017021141.1
XP_016876630.1
12
1943
XM_017021142.1
XP_016876631.1
12
1943
XM_017021143.1
XP_016876632.1
12
1943
XM_017021144.1
XP_016876633.1
12
1943
XM_017021145.1
XP_016876634.1
12
1943
GenBank mRNA
AB020691.1
-
7
993
AB511280.1
-
12
1474
AK022988.1
-
7
956
AY596970.1
-
12
1865
AY596971.1
-
12
1865
BC016305.1
-
7
1009
BC150596.1
-
12
1573
9056966
Gene Symbol: RALGAPA1P1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_104269.1
1
1904
Back To Top
More Information
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
regulation of small GTPase mediated signal transduction
activation of GTPase activity
GTPase activator activity
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0