Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RALGAPA1 SIRNA Assays ›
          Gene Symbol:
          RALGAPA1 RALGAPA1P1
          Gene Name:
          Ral GTPase activating protein catalytic alpha subunit 1
          Ral GTPase activating protein catalytic alpha subunit 1 pseudogene 1
          Gene Aliases:
          GARNL1, GRIPE, RalGAPalpha1, TULIP1, p240
          GARNL2, GARNL2P, RALGAPA1P, bA235C23.1
          Chromosome Location:
          Chr.14: 35538352 - 35809304 on Build GRCh38
          Chromosome Location:
          Chr.9: 105519824 - 105527319 on Build GRCh38
          UniGene ID:
          Hs.113150
          Hs.113150
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s48469

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RALGAPA1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001283043.1 NP_001269972.1 16 2636
          NM_001283044.1 NP_001269973.1 16 2636
          NM_014990.1 NP_055805.1 16 2636
          NM_194301.2 NP_919277.2 16 2636
          XM_005267491.3 XP_005267548.1 16 2714
          XM_005267492.3 XP_005267549.1 16 2714
          XM_005267493.3 XP_005267550.1 16 2714
          XM_006720098.2 XP_006720161.1 16 2714
          XM_006720099.2 XP_006720162.1 16 2714
          XM_006720100.3 XP_006720163.1 16 2714
          XM_006720101.2 XP_006720164.1 16 2714
          XM_006720102.3 XP_006720165.1 16 2420
          XM_006720103.2 XP_006720166.1 16 2714
          XM_006720104.3 XP_006720167.1 16 2714
          XM_011536615.2 XP_011534917.1 16 2714
          XM_011536616.2 XP_011534918.1 16 2714
          XM_017021141.1 XP_016876630.1 16 2714
          XM_017021142.1 XP_016876631.1 16 2714
          XM_017021143.1 XP_016876632.1 16 2714
          XM_017021144.1 XP_016876633.1 16 2714
          XM_017021145.1 XP_016876634.1 16 2714
          GenBank mRNA AB020691.1 - 11 1764
          AB511280.1 - 16 2245
          AK022988.1 - 11 1727
          AY596970.1 - 16 2636
          AY596971.1 - 16 2636
          BC150596.1 - 16 2344 9056966
          Gene Symbol: RALGAPA1P1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_104269.1 1 2675

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          regulation of small GTPase mediated signal transduction
          activation of GTPase activity
          GTPase activator activity
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0