Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMCO4 SIRNA Assays ›
Gene Symbol:
TMCO4
Gene Name:
transmembrane and coiled-coil domains 4
Gene Aliases:
-
Chromosome Location:
Chr.1: 19682213 - 19800385 on Build GRCh38
UniGene ID:
Hs.656313
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s48613
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMCO4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_181719.4
NP_859070.3
5, 6
604
XM_005245822.4
XP_005245879.1
5, 6
620
XM_005245824.4
XP_005245881.1
5, 6
684
XM_005245825.4
XP_005245882.1
4, 5
485
XM_005245827.3
XP_005245884.1
5, 6
620
XM_006710551.3
XP_006710614.1
5, 6
709
XM_011541179.2
XP_011539481.1
8, 9
1079
XM_011541180.2
XP_011539482.1
8, 9
1234
XM_011541181.1
XP_011539483.1
7, 8
908
XM_011541185.2
XP_011539487.1
5, 6
627
XM_011541186.2
XP_011539488.1
5, 6
724
XM_017000913.1
XP_016856402.1
5, 6
628
XM_017000914.1
XP_016856403.1
7, 8
1128
XM_017000915.1
XP_016856404.1
7, 8
850
XM_017000916.1
XP_016856405.1
5, 6
762
XM_017000917.1
XP_016856406.1
6, 7
977
XM_017000918.1
XP_016856407.1
7, 8
1174
XM_017000919.1
XP_016856408.1
6, 7
901
XM_017000920.1
XP_016856409.1
5, 6
1119
XM_017000921.1
XP_016856410.1
6, 7
926
GenBank mRNA
AK309310.1
-
5, 6
620
AK309900.1
-
3, 4
419
AL049846.1
-
2, 3
370
AL049847.1
-
2, 3
368
AX775873.1
-
4, 5
530
AX775875.1
-
4, 5
633
AX775877.1
-
4, 5
633
BC053600.1
-
3, 4
463
5744211
BX640987.1
-
5, 6
605
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-f7c8b:80/100.66.78.85:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0