Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MORN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173855.4 | NP_776254.3 | 1, 2 | 314 | ||
XM_005253869.4 | XP_005253926.1 | 2, 3 | 243 | |||
XM_011538213.2 | XP_011536515.1 | 2, 3 | 254 | |||
XM_011538214.2 | XP_011536516.1 | 1, 2 | 440 | |||
GenBank mRNA | AY177732.1 | - | 2, 3 | 225 | ||
BC057760.1 | - | 1, 2 | 314 | 30346478 |