Search
Search

F8| Gene Symbol: F8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000132.3 | NP_000123.1 | 24 | 6786 | ||
| NM_019863.2 | NP_063916.1 | 3 | 355 | |||
| GenBank mRNA | AK289947.1 | - | 3 | 285 | ||
| AK295164.1 | - | 12 | 1583 | |||
| AK313707.1 | - | 3 | 240 | |||
| AX746360.1 | - | 24 | 6615 | |||
| BC022513.1 | - | 3 | 274 | |||
| BC064380.1 | - | 3 | 334 | |||
| BC098389.1 | - | 3 | 353 | |||
| BC111967.1 | - | 3 | 243 | |||
| BC111969.1 | - | 3 | 242 | |||
| BI547023.1 | - | 3 | 392 | |||
| K01740.1 | - | 24 | 6765 | |||
| KT878539.1 | - | 1 | 36 | |||
| M14113.1 | - | 24 | 6786 | |||
| M90707.1 | - | 3 | 250 | |||
| X01179.1 | - | 24 | 6724 | |||