Search
Search

F8| Gene Symbol: F8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000132.3 | NP_000123.1 | 23 | 6626 | ||
| NM_019863.2 | NP_063916.1 | 2 | 195 | |||
| GenBank mRNA | AK289947.1 | - | 2 | 125 | ||
| AK295164.1 | - | 11 | 1423 | |||
| AK313707.1 | - | 2 | 80 | |||
| AX746360.1 | - | 23 | 6455 | |||
| BC022513.1 | - | 2 | 114 | |||
| BC064380.1 | - | 2 | 174 | |||
| BC098389.1 | - | 2 | 193 | |||
| BC111967.1 | - | 2 | 83 | |||
| BC111969.1 | - | 2 | 82 | |||
| K01740.1 | - | 23 | 6605 | |||
| M14113.1 | - | 23 | 6626 | |||
| M90707.1 | - | 2 | 90 | |||
| X01179.1 | - | 23 | 6564 | |||