Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012232.5 | NP_036364.2 | 2 | 2253 | ||
GenBank mRNA | AK000715.1 | - | 2 | 1739 | ||
AK074512.1 | - | 1454 | ||||
AK092406.1 | - | 4 | 1891 | |||
BC004295.1 | - | 1 | 1463 | |||
BC008849.1 | - | 2 | 1536 | 4140111 | ||
BC066123.1 | - | 2 | 2189 | 5269172 | ||
BC073759.1 | - | 2 | 2075 | 6572478 |