Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ENG SIRNA Assays ›
          Gene Symbol:
          ENG
          Gene Name:
          endoglin
          Gene Aliases:
          END, HHT1, ORW1
          Chromosome Location:
          Chr.9: 127815012 - 127854773 on Build GRCh38
          UniGene ID:
          Hs.76753
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s501306

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ENG
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000118.3 NP_000109.1 8 1532
          NM_001114753.2 NP_001108225.1 8 1532
          NM_001278138.1 NP_001265067.1 8 1272
          GenBank mRNA AK222669.1 - 8 1445
          AK223044.1 - 1 95
          AK290389.1 - 8 1416
          AK300156.1 - 3 249
          AK301171.1 - 8 1272
          AK302455.1 - 5 644
          BC014271.2 - 8 1266 3843174
          BT006872.1 - 8 1114
          J05481.1 - 8 1075
          X72012.1 - 8 1395

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          chronological cell aging
          patterning of blood vessels
          vasculogenesis
          positive regulation of protein phosphorylation
          heart looping
          sprouting angiogenesis
          endocardial cushion morphogenesis
          cardiac ventricle morphogenesis
          cardiac atrium morphogenesis
          ventricular trabecula myocardium morphogenesis
          cell migration involved in endocardial cushion formation
          regulation of transcription, DNA-templated
          cell adhesion
          negative regulation of gene expression
          positive regulation of pathway-restricted SMAD protein phosphorylation
          regulation of transforming growth factor beta receptor signaling pathway
          central nervous system vasculogenesis
          negative regulation of cell migration
          BMP signaling pathway
          positive regulation of BMP signaling pathway
          negative regulation of protein autophosphorylation
          dorsal aorta morphogenesis
          positive regulation of angiogenesis
          positive regulation of transcription from RNA polymerase II promoter
          smooth muscle tissue development
          artery morphogenesis
          venous blood vessel morphogenesis
          endocardial cushion to mesenchymal transition
          positive regulation of epithelial to mesenchymal transition involved in endocardial cushion formation
          transmembrane signaling receptor activity
          transforming growth factor beta receptor, cytoplasmic mediator activity
          type II transforming growth factor beta receptor binding
          protein binding
          galactose binding
          glycosaminoglycan binding
          type I transforming growth factor beta receptor binding
          BMP binding
          protein homodimerization activity
          activin binding
          transforming growth factor beta binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-grtsk:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0