Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATP2C1 SIRNA Assays ›
Gene Symbol:
ATP2C1
Gene Name:
ATPase secretory pathway Ca2+ transporting 1
Gene Aliases:
ATP2C1A, BCPM, HHD, PMR1, SPCA1, hSPCA1
Chromosome Location:
Chr.3: 130850500 - 131016712 on Build GRCh38
UniGene ID:
Hs.584884
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s502429
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATP2C1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001485.2
NP_001001485.1
23, 24
2629
NM_001001486.1
NP_001001486.1
22, 23
2306
NM_001001487.1
NP_001001487.1
22, 23
2306
NM_001199179.1
NP_001186108.1
23, 24
2535
NM_001199180.1
NP_001186109.1
22, 23
2524
NM_001199181.1
NP_001186110.1
22, 23
2524
NM_001199182.1
NP_001186111.1
21, 22
2407
NM_001199183.1
NP_001186112.1
22, 23
2397
NM_001199184.1
NP_001186113.1
22, 23
2397
NM_001199185.1
NP_001186114.1
22, 23
2448
NM_014382.3
NP_055197.2
22, 23
2448
XM_005247354.2
XP_005247411.1
22, 23
2549
XM_005247355.2
XP_005247412.1
23, 24
2676
XM_005247356.2
XP_005247413.1
23, 24
2429
XM_005247358.2
XP_005247415.1
22, 23
2253
XM_011512686.2
XP_011510988.1
13, 14
1358
XM_017006164.1
XP_016861653.1
23, 24
2337
GenBank mRNA
AB037768.1
-
23, 24
2536
AB209265.1
-
11, 12
2296
AF181120.1
-
22, 23
2361
AF181121.1
-
22, 23
2361
AF189723.2
-
22, 23
2280
AF225981.1
-
22, 23
2280
AJ010953.1
-
8, 9
858
AK001684.1
-
10, 11
1056
AK074692.1
-
20, 21
1987
AK296470.1
-
22, 23
2451
AK299945.1
-
21, 22
2163
AK304175.1
-
22, 23
2253
AK304374.1
-
22, 23
2280
AK314342.1
-
22, 23
2310
AY268374.1
-
22, 23
2255
AY268375.1
-
22, 23
2255
BC028139.1
-
22, 23
2318
5215224
Back To Top
More Information
Panther Classification:
Molecular Function -
primary active transporter
Gene Ontology Categories:
Function(s)
Process(es)
calcium ion transport
cellular calcium ion homeostasis
signal transduction
epidermis development
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
actin cytoskeleton reorganization
Golgi calcium ion homeostasis
Golgi calcium ion transport
ion transmembrane transport
positive regulation of I-kappaB kinase/NF-kappaB signaling
calcium ion transmembrane transport
signal transducer activity
calcium-transporting ATPase activity
ATP binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-bn9m5:80/10.239.167.51:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0