Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATP2C1 SIRNA Assays ›
Gene Symbol:
ATP2C1
Gene Name:
ATPase secretory pathway Ca2+ transporting 1
Gene Aliases:
ATP2C1A, BCPM, HHD, PMR1, SPCA1, hSPCA1
Chromosome Location:
Chr.3: 130850500 - 131016712 on Build GRCh38
UniGene ID:
Hs.584884
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s502430
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATP2C1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001485.2
NP_001001485.1
3
587
NM_001001486.1
NP_001001486.1
2
264
NM_001001487.1
NP_001001487.1
2
264
NM_001199179.1
NP_001186108.1
3
493
NM_001199180.1
NP_001186109.1
2
482
NM_001199181.1
NP_001186110.1
2
482
NM_001199182.1
NP_001186111.1
2
482
NM_001199183.1
NP_001186112.1
2
355
NM_001199184.1
NP_001186113.1
2
355
NM_001199185.1
NP_001186114.1
2
406
NM_014382.3
NP_055197.2
2
406
XM_005247354.2
XP_005247411.1
2
507
XM_005247355.2
XP_005247412.1
3
634
XM_005247356.2
XP_005247413.1
3
387
XM_005247358.2
XP_005247415.1
2
211
XM_017006164.1
XP_016861653.1
3
295
GenBank mRNA
AB037768.1
-
3
494
AF181120.1
-
2
319
AF181121.1
-
2
319
AF189723.2
-
2
238
AF225981.1
-
2
238
AK296470.1
-
2
409
AK299945.1
-
2
238
AK304175.1
-
2
211
AK304374.1
-
2
238
AK314342.1
-
2
268
AY268374.1
-
2
213
AY268375.1
-
2
213
BC028139.1
-
2
276
5215224
DC343471.1
-
2
406
DC345627.1
-
2
355
DC422788.1
-
2
482
Back To Top
More Information
Panther Classification:
Molecular Function -
primary active transporter
Gene Ontology Categories:
Function(s)
Process(es)
calcium ion transport
cellular calcium ion homeostasis
signal transduction
epidermis development
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
actin cytoskeleton reorganization
Golgi calcium ion homeostasis
Golgi calcium ion transport
ion transmembrane transport
positive regulation of I-kappaB kinase/NF-kappaB signaling
calcium ion transmembrane transport
signal transducer activity
calcium-transporting ATPase activity
ATP binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0