Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPFIA1 SIRNA Assays ›
Gene Symbol:
PPFIA1
Gene Name:
PTPRF interacting protein alpha 1
Gene Aliases:
LIP.1, LIP1, LIPRIN
Chromosome Location:
Chr.11: 70270687 - 70384501 on Build GRCh38
UniGene ID:
Hs.530749
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s505
Validated
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
7,898.00
Your Price
Online offer:
7898.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPFIA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_003626.3
NP_003617.1
17
2407
NM_177423.2
NP_803172.1
17
2407
NR_045286.1
18
2418
XM_006718716.3
XP_006718779.1
20
2642
XM_011545306.2
XP_011543608.1
20
2514
XM_011545307.2
XP_011543609.1
20
2642
XM_011545308.2
XP_011543610.1
19
2612
XM_011545309.2
XP_011543611.1
20
2642
XM_011545310.2
XP_011543612.1
19
2567
XM_011545311.2
XP_011543613.1
20
2642
XM_011545312.2
XP_011543614.1
20
2642
XM_011545313.2
XP_011543615.1
19
2525
XM_011545314.2
XP_011543616.1
18
2495
XM_011545315.2
XP_011543617.1
18
2450
XM_011545316.2
XP_011543618.1
19
2525
XM_011545317.2
XP_011543619.1
18
2495
XM_011545318.2
XP_011543620.1
5
747
XM_011545319.1
XP_011543621.1
18
2450
XM_017018448.1
XP_016873937.1
19
2612
XM_017018449.1
XP_016873938.1
18
2450
XM_017018450.1
XP_016873939.1
20
2642
GenBank mRNA
AB209314.1
-
18
2345
BC034046.1
-
17
2370
4794300
BX648037.1
-
14
1614
D49354.1
-
2
179
U22815.1
-
17
2421
U22816.1
-
17
2421
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00170349_m1
Gene Ontology Categories:
Function(s)
Process(es)
cell-matrix adhesion
signal transduction
neurotransmitter secretion
glutamate secretion
negative regulation of stress fiber assembly
negative regulation of establishment of protein localization to plasma membrane
signal transducer activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline