Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TMEM52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178545.3 | NP_848640.1 | 4 | 349 | ||
GenBank mRNA | AJ278736.1 | - | 4 | 363 | ||
AY358544.1 | - | 1 | 509 | |||
CN263898.1 | - | 4 | 361 |