Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: KY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178554.4 | NP_848649.3 | 2, 3 | 255 | ||
XM_017006287.1 | XP_016861776.1 | 2, 3 | 5286 | |||
XM_017006288.1 | XP_016861777.1 | 2, 3 | 5232 | |||
GenBank mRNA | AK090526.1 | - | 2, 3 | 255 | ||
AK294500.1 | - | 2, 3 | 256 | |||
BC136349.1 | - | 2, 3 | 216 | |||
DA238247.1 | - | 2, 3 | 255 |