Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FCGR3B SIRNA Assays ›
          Gene Symbol:
          FCGR3B FCGR3A
          Gene Name:
          Fc fragment of IgG receptor IIIb
          Fc fragment of IgG receptor IIIa
          Gene Aliases:
          CD16, CD16b, FCG3, FCGR3, FCR-10, FCRIII, FCRIIIb
          CD16, CD16A, FCG3, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3, IMD20
          Chromosome Location:
          Chr.1: 161623196 - 161631963 on Build GRCh38
          Chromosome Location:
          Chr.1: 161541759 - 161550623 on Build GRCh38
          UniGene ID:
          Hs.372679
          Hs.372679
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s5079

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FCGR3B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000570.4 NP_000561.3 5 534
          NM_001244753.1 NP_001231682.1 4 780
          NM_001271035.1 NP_001257964.1 4 777
          NM_001271036.1 NP_001257965.1 4 646
          GenBank mRNA AB025256.1 - 4 445
          AJ581669.1 - 4 445
          AK313219.1 - 4 526
          AK316565.1 - 4 466
          BC128562.1 - 4 429 40126686
          DA672751.1 - 5 534
          DA673839.1 - 4 543
          J04162.1 - 4 429
          M24854.1 - 4 510
          X07934.1 - 4 445
          X16863.1 - 4 445
          Gene Symbol: FCGR3A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001127593.1 NP_001121065.1 5 567
          NM_001127595.1 NP_001121067.1 5 549
          NM_001127596.1 NP_001121068.1 5 546
          GenBank mRNA AK223268.1 - 4 428
          AK223295.1 - 4 440
          AK302126.1 - 4 779
          BC017865.1 - 4 460 4690249
          BC033678.1 - 4 435 5180561
          BC036723.1 - 4 704 5206097
          BI768140.1 - 4 704
          DA674960.1 - 5 526
          DA944383.1 - 4 473
          KU178060.1 - 4 520
          M24853.1 - 2 322
          S76824.1 - 2 349
          X52645.1 - 4 445

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of defense response to virus by host
          immune response
          mitophagy in response to mitochondrial depolarization
          xenophagy
          Fc-gamma receptor signaling pathway involved in phagocytosis
          regulation of immune response
          IgG binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline