Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FKTN SIRNA Assays ›
          Gene Symbol:
          FKTN
          Gene Name:
          fukutin
          Gene Aliases:
          CMD1X, FCMD, LGMD2M, MDDGA4, MDDGB4, MDDGC4
          Chromosome Location:
          Chr.9: 105558117 - 105655950 on Build GRCh38
          UniGene ID:
          Hs.55777
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s5086

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FKTN
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001079802.1 NP_001073270.1 8, 9 1118
          NM_001198963.1 NP_001185892.1 8, 9 1118
          NM_006731.2 NP_006722.2 7, 8 1026
          XM_011518368.2 XP_011516670.1 9, 10 1266
          XM_011518369.2 XP_011516671.1 8, 9 1116
          XM_011518370.2 XP_011516672.1 9, 10 2802
          XM_011518371.2 XP_011516673.1 9, 10 1301
          XM_011518373.2 XP_011516675.1 9, 10 1212
          XM_011518374.2 XP_011516676.1 7, 8 1030
          XM_011518375.2 XP_011516677.1 7, 8 1345
          XM_011518376.2 XP_011516678.1 7, 8 1168
          XM_011518378.2 XP_011516680.1 9, 10 1266
          XM_011518379.2 XP_011516681.1 7, 8 912
          XM_011518381.2 XP_011516683.1 8, 9 1101
          XM_011518389.2 XP_011516691.1 9, 10 1266
          XM_011518390.2 XP_011516692.1 5, 6 707
          XM_017014462.1 XP_016869951.1 10, 11 1400
          XM_017014463.1 XP_016869952.1 7, 8 1489
          XM_017014464.1 XP_016869953.1 9, 10 1266
          XM_017014465.1 XP_016869954.1 8, 9 1116
          XM_017014466.1 XP_016869955.1 9, 10 1266
          XM_017014467.1 XP_016869956.1 8, 9 1525
          XM_017014468.1 XP_016869957.1 7, 8 1408
          XM_017014469.1 XP_016869958.1 9, 10 1266
          XM_017014470.1 XP_016869959.1 9, 10 1266
          XM_017014471.1 XP_016869960.1 10, 11 1364
          XM_017014472.1 XP_016869961.1 10, 11 1390
          XM_017014473.1 XP_016869962.1 5, 6 3268
          XM_017014474.1 XP_016869963.1 7, 8 1029
          XM_017014476.1 XP_016869965.1 10, 11 1384
          XM_017014477.1 XP_016869966.1 9, 10 1249
          XM_017014478.1 XP_016869967.1 11, 12 1451
          XM_017014479.1 XP_016869968.1 8, 9 1148
          GenBank mRNA AB008226.1 - 7, 8 1013
          AB609007.1 - 1013
          AK300840.1 - 8, 9 1116
          BC101808.1 - 6, 7 928 8069314
          BC112038.1 - 6, 7 928 8327506
          BC117699.1 - 6, 7 934 40073633
          BC117700.1 - 8, 9 1176 40073634

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          nervous system development
          muscle organ development
          negative regulation of cell proliferation
          protein O-linked mannosylation
          negative regulation of JNK cascade
          regulation of protein glycosylation
          transferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0