Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NMNAT3 SIRNA Assays ›
          Gene Symbol:
          NMNAT3
          Gene Name:
          nicotinamide nucleotide adenylyltransferase 3
          Gene Aliases:
          FKSG76, PNAT-3, PNAT3
          Chromosome Location:
          Chr.3: 139560180 - 139678050 on Build GRCh38
          UniGene ID:
          Hs.208673
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s51394

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NMNAT3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001200047.2 NP_001186976.1 4 774
          NM_001320510.1 NP_001307439.1 7 1323
          NM_001320511.1 NP_001307440.1 7 1075
          NM_001320512.1 NP_001307441.1 6 1041
          NM_001320513.1 NP_001307442.1 7 1453
          NM_178177.4 NP_835471.1 5 892
          XM_011512790.1 XP_011511092.1 4 467
          XM_011512791.2 XP_011511093.1 4 766
          XM_017006328.1 XP_016861817.1 8 1340
          XM_017006329.1 XP_016861818.1 8 1337
          XM_017006330.1 XP_016861819.1 7 1300
          XM_017006331.1 XP_016861820.1 8 1413
          XM_017006332.1 XP_016861821.1 8 5354
          XM_017006333.1 XP_016861822.1 6 1203
          XM_017006334.1 XP_016861823.1 6 1200
          XM_017006335.1 XP_016861824.1 7 1052
          XM_017006336.1 XP_016861825.1 6 1012
          XM_017006337.1 XP_016861826.1 5 915
          XM_017006338.1 XP_016861827.1 4 467
          XM_017006339.1 XP_016861828.1 6 1155
          XM_017006340.1 XP_016861829.1 5 1124
          XM_017006341.1 XP_016861830.1 7 1083
          XM_017006342.1 XP_016861831.1 8 1189
          XM_017006343.1 XP_016861832.1 7 1149
          XM_017006344.1 XP_016861833.1 6 1049
          GenBank mRNA AF345564.1 - 4 591
          AK123208.1 - 4 689
          AK127477.1 - 2 2877
          BC034374.1 - 5 840 5185621
          BC036218.1 - 7 1420 5270916
          BX649063.1 - 2 1722

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          nucleotidyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          NAD biosynthetic process
          response to wounding
          NAD metabolic process
          'de novo' NAD biosynthetic process from aspartate
          nicotinamide-nucleotide adenylyltransferase activity
          nicotinate-nucleotide adenylyltransferase activity
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline