Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: B3GNT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198540.2 | NP_940942.1 | 3 | 1827 | ||
XM_011526934.2 | XP_011525236.1 | 2 | 3146 | |||
GenBank mRNA | AY277592.2 | - | 3 | 1827 |