Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172643.1 | NP_001166114.1 | 10 | 668 | ||
NM_001172644.1 | NP_001166115.1 | 9 | 1029 | |||
NM_001172645.1 | NP_001166116.1 | 9 | 1050 | |||
NM_001271934.1 | NP_001258863.1 | 11 | 1199 | |||
NM_001271935.1 | NP_001258864.1 | 8 | 499 | |||
NM_001272050.1 | NP_001258979.1 | 10 | 878 | |||
NM_004614.4 | NP_004605.4 | 10 | 1104 | |||
NR_073520.1 | 9 | 2032 | ||||
GenBank mRNA | AF521891.1 | - | 10 | 1104 | ||
AK026021.1 | - | 4 | 244 | |||
AK295808.1 | - | 9 | 739 | |||
AK302976.1 | - | 10 | 882 | |||
AK303590.1 | - | 9 | 828 | |||
AK316226.1 | - | 9 | 739 | |||
BC113928.1 | - | 8 | 478 | |||
BC134344.1 | - | 10 | 647 | |||
BC142970.1 | - | 11 | 1000 | 8860391 | ||
U77088.1 | - | 10 | 668 | |||
U80628.1 | - | 9 | 1419 | |||
Y10498.3 | - | 10 | 872 |