Search
Search

BEND4| Gene Symbol: BEND4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159547.1 | NP_001153019.1 | 5 | 1859 | ||
| NM_207406.3 | NP_997289.2 | 5 | 1859 | |||
| XM_017008185.1 | XP_016863674.1 | 4 | 1516 | |||
| GenBank mRNA | AB126828.1 | - | 5 | 1859 | ||
| AB126829.1 | - | 5 | 1859 | |||
| AK125953.1 | - | 5 | 1045 | |||
| BC128592.1 | - | 5 | 944 | 40127911 | ||
| BC128593.1 | - | 5 | 944 | 40127915 | ||